Assay Details
Gene Symbol: mlt-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078110.4 | NP_510511.1 | 5 - 6 | 515 | 67 | |
NM_078111.4 | NP_510512.1 | 5 - 6 | 457 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
