Assay Details
| Gene Symbol: ocr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078119.7 | NP_510520.4 | 2 - 3 | 436 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ocr-3| Gene Symbol: ocr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078119.7 | NP_510520.4 | 2 - 3 | 436 | 57 | |
Set Membership: |
|