Assay Details
Gene Symbol: gcy-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078156.8 | NP_510557.3 | 9 - 10 | 1165 | 68 | |
GenBank mRNA | AY652945.1 | - | 9 - 10 | 1015 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
