Assay Details
| Gene Symbol: tbh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270335.2 | NP_001257264.1 | 2 - 3 | 289 | 75 | |
| NM_001270336.3 | NP_001257265.1 | 1 - 2 | 86 | 75 | ||
| NM_001313436.5 | NP_001300365.1 | 3 - 4 | 236 | 75 | ||
More Information
Set Membership: |
|

tbh-1
Genomic Map