Assay Details
| Gene Symbol: tbh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270335.2 | NP_001257264.1 | 5 - 6 | 686 | 114 | |
| NM_001270336.3 | NP_001257265.1 | 4 - 5 | 483 | 114 | ||
| NM_001313436.5 | NP_001300365.1 | 6 - 7 | 633 | 114 | ||
More Information
Set Membership: |
|

tbh-1
Genomic Map