Assay Details
| Gene Symbol: tbh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270335.2 | NP_001257264.1 | 8 - 9 | 1690 | 159 | |
| NM_001270336.3 | NP_001257265.1 | 7 - 8 | 1487 | 159 | ||
| NM_001313436.5 | NP_001300365.1 | 9 - 10 | 1637 | 159 | ||
More Information
Set Membership: |
|

tbh-1
Genomic Map