Assay Details
Gene Symbol: gpn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078181.4 | NP_510582.1 | 1 - 2 | 223 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
