Assay Details
Gene Symbol: mrp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373606.3 | NP_001359552.1 | 18 - 19 | 3639 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
