Assay Details
| Gene Symbol: mboa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078222.6 | NP_510623.1 | 7 - 8 | 1288 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mboa-1| Gene Symbol: mboa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078222.6 | NP_510623.1 | 7 - 8 | 1288 | 67 | |
Set Membership: |
|