Assay Details
Gene Symbol: sgk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047834.4 | NP_001041299.1 | 10 - 11 | 1294 | 119 | |
NM_001381177.1 | NP_001367134.1 | 10 - 11 | 1428 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
