Assay Details
| Gene Symbol: sdc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373609.3 | NP_001360585.1 | 5 - 6 | 1172 | 108 | |
| NM_078249.7 | NP_510650.2 | 5 - 6 | 1160 | 108 | ||
More Information
Set Membership: |
|

sdc-1
Genomic Map