Assay Details
Gene Symbol: sdc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373609.3 | NP_001360585.1 | 6 - 7 | 1357 | 133 | |
NM_078249.7 | NP_510650.2 | 6 - 7 | 1345 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
