Assay Details
Gene Symbol: taf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270343.3 | NP_001257272.1 | 7 - 8 | 2302 | 82 | |
NM_001270344.3 | NP_001257273.1 | 1 - 2 | 211 | 82 | ||
NM_001347279.5 | NP_001334235.1 | 6 - 7 | 2008 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
