Assay Details
Gene Symbol: taf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270343.3 | NP_001257272.1 | 8 - 9 | 2493 | 83 | |
NM_001270344.3 | NP_001257273.1 | 2 - 3 | 402 | 83 | ||
NM_001347279.5 | NP_001334235.1 | 7 - 8 | 2199 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
