Assay Details
| Gene Symbol: mbl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136459.3 | NP_001129931.1 | 2 - 3 | 255 | 66 | |
| NM_001136460.5 | NP_001129932.1 | 4 - 5 | 512 | 66 | ||
| NM_001270352.4 | NP_001257281.1 | 4 - 5 | 422 | 66 | ||
| NM_001373620.2 | NP_001360647.1 | 4 - 5 | 458 | 66 | ||
| NM_001373621.2 | NP_001360648.1 | 4 - 5 | 458 | 66 | ||
| NM_001373622.1 | NP_001360649.1 | 4 - 5 | 422 | 66 | ||
| NM_001373623.3 | NP_001360650.1 | 4 - 5 | 413 | 66 | ||
More Information
Set Membership: |
|

mbl-1
Genomic Map