Assay Details
| Gene Symbol: gcy-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373625.3 | NP_001359843.1 | 9 - 10 | 1136 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-11| Gene Symbol: gcy-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373625.3 | NP_001359843.1 | 9 - 10 | 1136 | 69 | |
Set Membership: |
|