Assay Details
| Gene Symbol: csk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306233.3 | NP_001293162.1 | 4 - 5 | 502 | 89 | |
| NM_001380379.1 | NP_001367885.1 | 3 - 4 | 417 | 89 | ||
| GenBank mRNA | AB096875.1 | - | 3 - 4 | 404 | 89 | |
More Information
Set Membership: |
|

csk-1
Genomic Map