Assay Details
| Gene Symbol: wwp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306342.3 | NP_001293271.1 | 4 - 5 | 1009 | 66 | |
| NM_001306343.3 | NP_001293272.1 | 4 - 5 | 1003 | 66 | ||
| NM_170794.7 | NP_740775.1 | 8 - 9 | 1886 | 66 | ||
| NM_171831.4 | NP_740776.1 | 8 - 9 | 1880 | 66 | ||
More Information
Set Membership: |
|

wwp-1
Genomic Map