Assay Details
| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059957.5 | NP_492358.3 | 17 - 18 | 2639 | 113 | |
| NM_170904.3 | NP_740896.2 | 15 - 16 | 2528 | 113 | ||
| NM_170905.3 | NP_740897.1 | 9 - 10 | 1538 | 113 | ||
| NM_170906.6 | NP_740898.1 | 3 - 4 | 1064 | 113 | ||
More Information
Set Membership: |
|

cle-1
Genomic Map