Assay Details
| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059957.5 | NP_492358.3 | 19 - 20 | 2966 | 84 | |
| NM_170904.3 | NP_740896.2 | 17 - 18 | 2855 | 84 | ||
| NM_170905.3 | NP_740897.1 | 11 - 12 | 1865 | 84 | ||
| NM_170906.6 | NP_740898.1 | 5 - 6 | 1391 | 84 | ||
More Information
Set Membership: |
|

cle-1
Genomic Map