Assay Details
| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059957.5 | NP_492358.3 | 22 - 23 | 3452 | 110 | |
| NM_170904.3 | NP_740896.2 | 20 - 21 | 3341 | 110 | ||
| NM_170905.3 | NP_740897.1 | 14 - 15 | 2351 | 110 | ||
| NM_170906.6 | NP_740898.1 | 8 - 9 | 1877 | 110 | ||
More Information
Set Membership: |
|

cle-1
Genomic Map