Assay Details
| Gene Symbol: dut-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026382.6 | NP_001021553.1 | 2 - 3 | 506 | 63 | |
| GenBank mRNA | U96695.1 | - | 2 - 3 | 516 | 63 | |
More Information
Set Membership: |
|

dut-1
Genomic Map