Assay Details
Gene Symbol: kin-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381269.1 | NP_001368699.1 | 6 - 7 | 889 | 77 | |
NM_001381270.1 | NP_001367024.1 | 3 - 4 | 329 | 77 | ||
NM_001381271.1 | NP_001367023.1 | 3 - 4 | 215 | 77 | ||
NM_001381272.1 | NP_001366905.1 | 3 - 4 | 363 | 77 | ||
NM_001381273.1 | NP_001367025.1 | 3 - 4 | 296 | 77 | ||
NM_001381274.1 | NP_001367022.1 | 3 - 4 | 240 | 77 | ||
NM_001381275.2 | NP_001367021.1 | 2 - 3 | 1009 | 77 | ||
NM_061204.4 | NP_493605.1 | 3 - 4 | 363 | 77 | ||
NM_170958.5 | NP_740954.1 | 5 - 6 | 492 | 77 | ||
NM_170960.6 | NP_740956.1 | 3 - 4 | 267 | 77 | ||
NM_170962.5 | NP_740958.1 | 3 - 4 | 434 | 77 | ||
NM_170964.6 | NP_740960.1 | 3 - 4 | 439 | 77 | ||
NM_170965.3 | NP_740961.1 | 3 - 4 | 449 | 77 | ||
NM_170966.3 | NP_740962.1 | 3 - 4 | 569 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
