Assay Details
Gene Symbol: kin-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381269.1 | NP_001368699.1 | 5 - 6 | 762 | 86 | |
NM_001381270.1 | NP_001367024.1 | 2 - 3 | 202 | 86 | ||
NM_001381271.1 | NP_001367023.1 | 2 - 3 | 88 | 86 | ||
NM_001381272.1 | NP_001366905.1 | 2 - 3 | 236 | 86 | ||
NM_001381273.1 | NP_001367025.1 | 2 - 3 | 169 | 86 | ||
NM_001381274.1 | NP_001367022.1 | 2 - 3 | 113 | 86 | ||
NM_061204.4 | NP_493605.1 | 2 - 3 | 236 | 86 | ||
NM_170958.5 | NP_740954.1 | 4 - 5 | 365 | 86 | ||
NM_170960.6 | NP_740956.1 | 2 - 3 | 140 | 86 | ||
NM_170962.5 | NP_740958.1 | 2 - 3 | 307 | 86 | ||
NM_170964.6 | NP_740960.1 | 2 - 3 | 312 | 86 | ||
NM_170965.3 | NP_740961.1 | 2 - 3 | 322 | 86 | ||
NM_170966.3 | NP_740962.1 | 2 - 3 | 442 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
