Assay Details
Gene Symbol: kin-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170964.6 | NP_740960.1 | 1 - 2 | 245 | 85 | |
NM_170965.3 | NP_740961.1 | 1 - 2 | 255 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
