Assay Details
Gene Symbol: kin-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381275.2 | NP_001367021.1 | 1 - 2 | 881 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
