Assay Details
| Gene Symbol: eff-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026816.6 | NP_001021987.1 | 7 - 8 | 1860 | 100 | |
| NR_136365.1 | 8 - 9 | 1804 | 100 | |||
| GenBank mRNA | AF480492.1 | - | 7 - 8 | 1856 | 100 | |
More Information
Set Membership: |
|

eff-1
Genomic Map