Assay Details
| Gene Symbol: pals-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384001.1 | 1 - 2 | 106 | 129 | ||
| NM_171857.5 | NP_741063.1 | 1 - 2 | 93 | 129 | ||
| Gene Symbol: pals-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064745.5 | NP_497146.2 | 10 - 11 | 1205 | 129 | |
More Information
Set Membership: |
|

pals-23
Genomic Map