Assay Details
Gene Symbol: pll-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171059.6 | NP_741067.1 | 1 - 2 | 84 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
