Assay Details
Gene Symbol: pll-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171059.6 | NP_741067.1 | 7 - 8 | 922 | 67 | |
NM_171060.5 | NP_741068.1 | 7 - 8 | 950 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
