Assay Details
| Gene Symbol: sax-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171117.5 | NP_741131.2 | 26 - 27 | 7487 | 143 | |
| NM_171118.6 | NP_741132.2 | 1 - 2 | 971 | 143 | ||
More Information
Set Membership: |
|

sax-2
Genomic Map