Assay Details
| Gene Symbol: sax-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171117.5 | NP_741131.2 | 27 - 28 | 7828 | 64 | |
| NM_171118.6 | NP_741132.2 | 2 - 3 | 1312 | 64 | ||
More Information
Set Membership: |
|

sax-2
Genomic Map