Assay Details
| Gene Symbol: sax-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171117.5 | NP_741131.2 | 9 - 10 | 1370 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sax-2| Gene Symbol: sax-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171117.5 | NP_741131.2 | 9 - 10 | 1370 | 131 | |
Set Membership: |
|