Assay Details
| Gene Symbol: swp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025951.3 | NP_001021122.1 | 6 - 7 | 2087 | 65 | |
| NM_001379724.2 | NP_001367326.1 | 7 - 8 | 2130 | 65 | ||
| NM_001382877.1 | 7 - 8 | 2138 | 65 | |||
| NM_001382878.1 | 7 - 8 | 2253 | 65 | |||
| NM_171132.4 | NP_741149.1 | 5 - 6 | 2081 | 65 | ||
| GenBank mRNA | U06932.1 | - | 5 - 6 | 2081 | 65 | |
More Information
Set Membership: |
|

swp-1
Genomic Map