Assay Details
| Gene Symbol: aldo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026069.7 | NP_001021240.1 | 3 - 4 | 1026 | 62 | |
| GenBank mRNA | D83739.1 | - | 3 - 4 | 910 | 62 | |
More Information
Set Membership: |
|

aldo-2
Genomic Map