Assay Details
| Gene Symbol: esyt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379752.2 | NP_001366808.1 | 7 - 8 | 949 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

esyt-2| Gene Symbol: esyt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379752.2 | NP_001366808.1 | 7 - 8 | 949 | 119 | |
Set Membership: |
|