Assay Details
Gene Symbol: ten-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027552.5 | NP_001022723.2 | 15 - 16 | 8015 | 85 | |
NM_001268011.3 | NP_001254940.1 | 18 - 19 | 8409 | 85 | ||
NM_001268012.3 | NP_001254941.1 | 16 - 17 | 8187 | 85 | ||
NM_001379777.2 | NP_001367791.1 | 15 - 16 | 8021 | 85 | ||
GenBank mRNA | AB206835.1 | - | 15 - 16 | 8015 | 85 | |
AB206836.1 | - | 10 - 11 | 7521 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
