Assay Details
Gene Symbol: ten-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027552.5 | NP_001022723.2 | 11 - 12 | 3977 | 94 | |
NM_001268011.3 | NP_001254940.1 | 14 - 15 | 4371 | 94 | ||
NM_001268012.3 | NP_001254941.1 | 12 - 13 | 4149 | 94 | ||
NM_001379777.2 | NP_001367791.1 | 11 - 12 | 3983 | 94 | ||
GenBank mRNA | AB206835.1 | - | 11 - 12 | 3977 | 94 | |
AB206836.1 | - | 6 - 7 | 3483 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
