Assay Details
Gene Symbol: ten-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027552.5 | NP_001022723.2 | 13 - 14 | 5430 | 106 | |
NM_001268011.3 | NP_001254940.1 | 16 - 17 | 5824 | 106 | ||
NM_001268012.3 | NP_001254941.1 | 14 - 15 | 5602 | 106 | ||
NM_001379777.2 | NP_001367791.1 | 13 - 14 | 5436 | 106 | ||
GenBank mRNA | AB206835.1 | - | 13 - 14 | 5430 | 106 | |
AB206836.1 | - | 8 - 9 | 4936 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
