Assay Details
Gene Symbol: ten-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027552.5 | NP_001022723.2 | 14 - 15 | 7353 | 77 | |
NM_001268011.3 | NP_001254940.1 | 17 - 18 | 7747 | 77 | ||
NM_001268012.3 | NP_001254941.1 | 15 - 16 | 7525 | 77 | ||
NM_001379777.2 | NP_001367791.1 | 14 - 15 | 7359 | 77 | ||
NM_066086.8 | NP_498487.2 | 1 - 2 | 1903 | 77 | ||
GenBank mRNA | AB206835.1 | - | 14 - 15 | 7353 | 77 | |
AB206836.1 | - | 9 - 10 | 6859 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
