Assay Details
Gene Symbol: prk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276777.3 | NP_001263706.1 | 5 - 6 | 693 | 87 | |
NM_001377564.1 | NP_001364690.1 | 4 - 5 | 436 | 87 | ||
NM_001377565.2 | NP_001364691.1 | 4 - 5 | 436 | 87 | ||
NM_066127.4 | NP_498528.1 | 4 - 5 | 436 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
