Assay Details
| Gene Symbol: ran-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171198.7 | NP_741232.1 | 8 - 9 | 2700 | 77 | |
| NM_171884.5 | NP_741233.2 | 8 - 9 | 2804 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ran-2| Gene Symbol: ran-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171198.7 | NP_741232.1 | 8 - 9 | 2700 | 77 | |
| NM_171884.5 | NP_741233.2 | 8 - 9 | 2804 | 77 | ||
Set Membership: |
|