Assay Details
| Gene Symbol: aak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382945.1 | 2 - 3 | 487 | 118 | ||
| NM_171216.5 | NP_741254.3 | 2 - 3 | 488 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382945.1 transcripts.
Set Membership: |
|

aak-1
Genomic Map