Assay Details
| Gene Symbol: cdk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027576.5 | NP_001022747.1 | 4 - 5 | 740 | 102 | |
| GenBank mRNA | AF129109.1 | - | 1 - 2 | 212 | 102 | |
| X68384.1 | - | 196 | 102 | |||
More Information
Set Membership: |
|

cdk-1
Genomic Map