Assay Details
Gene Symbol: pad-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268140.3 | NP_001255069.1 | 2 - 3 | 518 | 111 | |
NM_001268141.3 | NP_001255070.1 | 2 - 3 | 515 | 111 | ||
GenBank mRNA | AF455271.1 | - | 2 - 3 | 509 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
