Assay Details
| Gene Symbol: rbm-39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383036.1 | 5 - 6 | 1044 | 96 | ||
| NM_067624.5 | NP_500025.1 | 5 - 6 | 1122 | 96 | ||
| NR_136395.1 | 6 - 7 | 1024 | 96 | |||
More Information
Set Membership: |
|

rbm-39
Genomic Map