Assay Details
| Gene Symbol: egl-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028052.5 | NP_001023223.1 | 3 - 4 | 663 | 63 | |
| NM_001307521.4 | NP_001294450.1 | 3 - 4 | 557 | 63 | ||
| NM_067740.6 | NP_500141.1 | 3 - 4 | 557 | 63 | ||
| NM_067741.5 | NP_500142.1 | 3 - 4 | 428 | 63 | ||
| NM_171279.6 | NP_741329.1 | 3 - 4 | 464 | 63 | ||
| Gene Symbol: ccm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380067.2 | NP_001367894.1 | 12 - 13 | 1455 | 63 | |
More Information
Set Membership: |
|

egl-4
Genomic Map