Assay Details
| Gene Symbol: egl-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028052.5 | NP_001023223.1 | 5 - 6 | 928 | 71 | |
| NM_001307521.4 | NP_001294450.1 | 5 - 6 | 822 | 71 | ||
| NM_067740.6 | NP_500141.1 | 5 - 6 | 822 | 71 | ||
| NM_067741.5 | NP_500142.1 | 5 - 6 | 693 | 71 | ||
| NM_171279.6 | NP_741329.1 | 5 - 6 | 729 | 71 | ||
| Gene Symbol: ccm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380067.2 | NP_001367894.1 | 14 - 15 | 1720 | 71 | |
More Information
Set Membership: |
|

egl-4
Genomic Map