Assay Details
| Gene Symbol: mgl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083266.5 | NP_001076735.3 | 12 - 13 | 2132 | 113 | |
| NM_001313057.3 | NP_001299986.1 | 6 - 7 | 832 | 113 | ||
| NM_001313058.3 | NP_001299987.1 | 6 - 7 | 832 | 113 | ||
| NM_001313059.3 | NP_001299988.1 | 2 - 3 | 238 | 113 | ||
| NM_001313060.3 | NP_001299989.1 | 2 - 3 | 238 | 113 | ||
| NM_171341.5 | NP_741400.2 | 12 - 13 | 2130 | 113 | ||
| NR_132461.1 | 12 - 13 | 2210 | 113 | |||
| NR_132462.1 | 12 - 13 | 2062 | 113 | |||
| NR_132463.1 | 12 - 13 | 2062 | 113 | |||
More Information
Set Membership: |
|

mgl-3
Genomic Map