Assay Details
Gene Symbol: mgl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313058.3 | NP_001299987.1 | 10 - 11 | 1392 | 74 | |
NM_001313060.3 | NP_001299989.1 | 6 - 7 | 798 | 74 | ||
NM_171341.5 | NP_741400.2 | 16 - 17 | 2690 | 74 | ||
NR_132461.1 | 16 - 17 | 2770 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
